Nickel aluminium alloy according to Raney

Supplier: BeanTown Chemical

Synonyms: Aluminum-nickel alloy

141125-2KG 141125-500G 141125-100G
BT141125-2KGEA 443.54 USD
BT141125-2KG BT141125-100G BT141125-500G
Nickel aluminium alloy according to Raney
Nickel aluminium alloy according to Raney

  • Powder

Formula: Ni:Al;₅₀:₅₀%
Melting Pt: 1350 °C
Density: 3.46 g/mL
MDL Number: MFCD00167392
CAS Number: 12635-27-7
EINECS: 231-111-4
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR