Cyclohexanecarboxaldehyde ≥97%

Supplier: BeanTown Chemical

141000-25G
BT141000-25GEA 71.39 USD
BT141000-25G
Cyclohexanecarboxaldehyde ≥97%
Cyclohexanecarboxaldehyde

  • Refractive index: 1.452
  • Air Sensitive
  • Liquid

Formula: C₇H₁₂O
Boiling Pt: 161…163 °C
Density: 0.926 g/mL
Flash Pt: 41 °C (106 °F)
MDL Number: MFCD00001457
CAS Number: 2043-61-0
EINECS: 218-057-7
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR