Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3% ACS

Synonyms: Ethylene glycol monomethyl ether

140585-250ML 140585-4L 140585-1L
BT140585-4LEA 155.77 USD
BT140585-4L BT140585-250ML BT140585-1L
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3% ACS
Ethylene glycol monomethyl ether (2-Methoxyethanol)

  • RTECS: KL5775000
  • Refractive index: 1.402
  • Liquid

Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124…125 °C
Melting Pt: –85 °C
Density: 0.965 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III

Order Now

Specification Test Results

Assay 99.3% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR