Vanadyl sulfate hydrate ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Vanadium oxide sulfate hydrate
, Basic vanadium (IV) sulfate hydrate
, Vanadium(IV) oxide sulfate hydrate
, Vanadium (IV) sulfate oxide hydrate
, Vanadium(IV) oxide sulphate hydrate
, Vanadium (IV) sulphate oxide hydrate
, Basic vanadium (IV) sulphate hydrate
, Vanadium oxide sulphate hydrate
139795-50G
139795-250G
139795-10G
BT139795-250GEA
395.21
USD
BT139795-250G
BT139795-50G
BT139795-10G
Vanadyl sulfate hydrate ≥99.9% (trace metals basis)
Vanadyl sulphate hydrate
- RTECS: YW1840000
- Lump
Formula:
O₅SV·H₂O MW: 163.00 g/mol (anhy) Melting Pt: 105 °C Density: 2.5 g/mL |
MDL Number:
MFCD00149933 CAS Number: 123334-20-3 EINECS: 248-652-7 UN: 2931 ADR: 6.1,II |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...