Methanol-[D₄] ≥99.8% (Isotopic) contains 0.05% TMS for NMR spectroscopy

Synonyms: Carbinol-D4Methyl alcohol-D4Methanol-[²H₄]Methyl-D3 alcohol-DWood alcohol-D4Methanol-d4Hydroxy methane-D4Tetradeuteromethanol

139690-10EACH 139690-2EACH
BT139690-2EACHEA 34.43 USD
BT139690-2EACH BT139690-10EA
Methanol-[D₄] ≥99.8% (Isotopic) contains 0.05% TMS for NMR spectroscopy
Methanol-[D4]

  • Refractive index: 1.326
  • Hygroscopic
  • Liquid, in prescored ampules

Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.4 °C
Melting Pt: –98 °C
Density: 0.888 g/mL
Flash Pt: 11 °C (54 °F)
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Assay 99.8%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR