Methanol ≥99.8% for biochemistry
Supplier: BeanTown Chemical
Synonyms:
Hydroxy methane
, Methyl hydroxide
, Methyl alcohol
, Methylol
, Carbinol
, Wood alcohol
139630-4X4L
139630-4L
BT139630-4X4LEA
244.66
USD
BT139630-4X4L
BT139630-4L
Methanol ≥99.8% for biochemistry
Methanol
- RTECS: PC1400000
- Refractive index: 1.329
- Liquid
Formula:
CH₃OH MW: 32.04 g/mol Boiling Pt: 64.7 °C Melting Pt: –98 °C Density: 0.791 g/mL Flash Pt: 9.7 °C (49.5 °F) |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II |
Specification Test Results
Assay | 99.8% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...