Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99%

Synonyms: Ethylene glycol monomethyl ether

139430-10L 139430-2.5L 139430-500ML
BT139430-10LEA 215.57 USD
BT139430-10L BT139430-500ML BT139430-2.5L
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99%
Ethylene glycol monomethyl ether (2-Methoxyethanol)

  • RTECS: KL5775000
  • Refractive index: 1.402
  • Liquid

Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124…125 °C
Melting Pt: –85 °C
Density: 0.965 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR