Hexacarbonylmolybdenum ≥98% may contain up to 1% Cr

Synonyms: Molybdenum carbonylMolybdenum hexacarbonylHexacarbonylmolybdenum(0)

139030-100G 139030-5G 139030-25G
BT139030-5GEA 48.26 USD
BT139030-5G BT139030-25G BT139030-100G
Hexacarbonylmolybdenum ≥98% may contain up to 1% Cr
Hexacarbonylmolybdenum

  • Crystalline

Formula: C₆MoO₆
MW: 264 g/mol
Melting Pt: 150 °C (dec.)
Density: 1.96 g/mL
MDL Number: MFCD00003466
CAS Number: 13939-06-5
EINECS: 237-713-3
UN: 3466
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR