Diphenylzinc ≥98%
Supplier: BeanTown Chemical
138660-5G
138660-1G
BT138660-1GEA
135.66
USD
BT138660-1G
BT138660-5G
Diphenylzinc ≥98%
Diphenylzinc
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₁₂H₁₀Zn Boiling Pt: 280…285 °C Melting Pt: 102…106 °C |
MDL Number:
MFCD0001407 CAS Number: 1078-58-6 EINECS: 214-082-2 ADR: 4.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...