4-Methyl-2-pentanone ≥99%

Synonyms: Methyl Isobutyl KetoneIsobutyl Methyl KetoneMethyl iso-Butyl Ketone

138565-2.5L 138565-500ML
BT138565-2.5LEA 93.5 USD
BT138565-2.5L BT138565-500ML
4-Methyl-2-pentanone ≥99%
4-Methyl-2-pentanone

  • RTECS: SA9275000
  • Refractive index: 1.395
  • Liquid

Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117…118 °C
Melting Pt: –80 °C
Density: 0.801 g/mL
Flash Pt: 14 °C (57 °F)
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/93/40/739340.jpg

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR