Iodine(V) oxide ≥98%
Supplier: BeanTown Chemical
Synonyms:
di-Iodine pentoxide
, di-Iodine pentaoxide
, Iodopentoxide
138355-100G
138355-25G
BT138355-100GEA
274.23
USD
BT138355-100G
BT138355-25G
Iodine(V) oxide ≥98%
Iodine (V) oxide
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
I₂O₅ MW: 333.81 g/mol Density: 5.08 g/mL |
MDL Number:
MFCD00011356 CAS Number: 12029-98-0 EINECS: 234-740-2 UN: 3085 ADR: 5.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...