Sodium perchlorate 98.0-102.0% ACS

Supplier: BeanTown Chemical

138240-100G 138240-500G
BT138240-100GEA 42.5 USD
BT138240-100G BT138240-500G
Sodium perchlorate 98.0-102.0% ACS
Sodium perchlorate

  • Hygroscopic
  • Crystalline/Flakes

Formula: ClNaO₄
MW: 122.44 g/mol
Melting Pt: 468 °C
Density: 2.499 g/mL
MDL Number: MFCD00003480
CAS Number: 7601-89-0
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98.0-102.0%
pH of 5% solution at 25°C 6.0-8.0
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.003%
Sulfate (SO4) ≤0.002%
Calcium (Ca) ≤0.02%
Iron (Fe) ≤0.0005%
Heavy metals (as Pb) ≤0.0005%
Potassium (K) ≤0.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR