Tricyclohexylphosphine oxide
Supplier: BeanTown Chemical
138230-25G
138230-1G
138230-5G
BT138230-25GEA
458.52
USD
BT138230-25G
BT138230-1G
BT138230-5G
Tricyclohexylphosphine oxide
Tricyclohexylphosphine oxide
- Crystalline
Formula:
C₁₈H₃₃OP MW: 296.42 g/mol Melting Pt: 155…157 °C |
MDL Number:
MFCD00014298 CAS Number: 13689-19-5 UN: 3464 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...