Perfluoro-tert-butanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Perfluoro-t-butanol1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanolPerfluoro-tert-butyl alcoholNonafluoro-tert-butyl alcohol

137865-5G 137865-25G 137865-1G
BT137865-25GEA 1172.72 USD
BT137865-25G BT137865-1G BT137865-5G
Perfluoro-tert-butanol ≥99%
Perfluoro-tert-butanol

  • RTECS: UB6452700
  • Refractive index: 1.300
  • Liquid

Formula: C₄HF₉O
MW: 236.04 g/mol
Boiling Pt: 45 °C
Melting Pt: –17 °C
Density: 1.693 g/mL
MDL Number: MFCD00042092
CAS Number: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR