Cyclopropanecarboxaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: Cyclopropanecarbaldehyde

137690-1G 137690-5G
BT137690-1GEA 37.6 USD
BT137690-1G BT137690-5G
Cyclopropanecarboxaldehyde ≥98%
Cyclopropanecarboxaldehyde

  • RTECS: GZ1005000
  • Refractive index: 1.430
  • Air Sensitive
  • Liquid

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 98…101 °C
Density: 0.938 g/mL
Flash Pt: 7 °C (45 °F)
MDL Number: MFCD00012261
CAS Number: 1489-69-6
ADR: 3(8),II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR