Benzyltrimethylammonium chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
N,N,N-Trimethyl-1-phenylmethanaminium chloride
137450-100G
137450-500G
137450-2.5KG
BT137450-2.5KGEA
326.14
USD
BT137450-2.5KG
BT137450-500G
BT137450-100G
Benzyltrimethylammonium chloride ≥97%
Benzyltrimethylammonium chloride
- RTECS: BO8400000
- Hygroscopic
- Crystalline
Formula:
C₁₀H₁₆ClN MW: 185.7 g/mol Melting Pt: 239 °C (dec.) |
MDL Number:
MFCD00011782 CAS Number: 56-93-9 EINECS: 200-300-3 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...