2-Butyne-1,4-diol ≥98%
Supplier: BeanTown Chemical
Synonyms:
But-2-yne-1,4-diol
, 1,4-Dihydroxy-2-butyne
137410-1KG
137410-250G
137410-5KG
BT137410-250GEA
28.24
USD
BT137410-250G
BT137410-1KG
BT137410-5KG
2-Butyne-1,4-diol ≥98%
2-Butyne-1,4-diol
- RTECS: ES0525000
- Solid
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 238 °C Melting Pt: 53…58 °C Flash Pt: 152 °C (306 °F) |
MDL Number:
MFCD00002915 CAS Number: 110-65-6 EINECS: 203-788-6 UN: 2716 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...