Zirconium tetrafluoride ≥98%

Supplier: BeanTown Chemical

Synonyms: Zirconium (IV) fluoride

136505-250G 136505-50G
BT136505-250GEA 360.32 USD
BT136505-250G BT136505-50G
Zirconium tetrafluoride ≥98%
Zirconium tetrafluoride

  • RTECS: ZH7875000
  • Refractive index: 1.59
  • Hygroscopic
  • Powder

Formula: F₄Zr
MW: 167.22 g/mol
Melting Pt: 640 °C
Density: 4.43 g/mL
MDL Number: MFCD00011307
CAS Number: 7783-64-4
EINECS: 232-018-1
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR