Sodium perrhenate ≥99.95% (trace metals basis)

Supplier: BeanTown Chemical

136415-5G 136415-1G 136415-250MG
BT136415-1GEA 47.89 USD
BT136415-1G BT136415-5G BT136415-250MG
Sodium perrhenate ≥99.95% (trace metals basis)
Sodium perrhenate

  • RTECS: WD3675000
  • Hygroscopic
  • Crystalline

Formula: NaO₄Re
MW: 273.19 g/mol
Melting Pt: 300 °C
Density: 5.390 g/mL
MDL Number: MFCD00014231
CAS Number: 13472-33-8
EINECS: 236-742-9
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR