4-(Methylamino)phenol sulfate 99.0-101.5% ACS

Synonyms: 4-(Methylamino)phenol hemisulfate saltMetol4-(Methylamino)phenol hemisulphatep-(Methylamino)phenol sulphate4-(Methylamino)phenol hemisulfate4-Methylaminophenolsulphatep-(Methylamino)phenol sulfate4-(Methylamino)phenol hemisulphate salt4-Methylaminophenolsulfate4-Methylaminophenol sulfate4-Methylaminophenol sulphate

136205-250G 136205-50G
BT136205-250GEA 104.96 USD
BT136205-250G BT136205-50G
4-(Methylamino)phenol sulfate 99.0-101.5% ACS
4-(Methylamino)phenol sulfate

  • RTECS: SL8650000
  • Air Sensitive, Light Sensitive
  • Powder

Formula: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
Melting Pt: 260 °C (dec.)
MDL Number: MFCD00013140
CAS Number: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.0-101.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR