Dimethyl acetylenedicarboxylate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dimethyl but-2-yne-1,4-dioate,
136000-100G
136000-500G
136000-25G
BT136000-25GEA
76.36
USD
BT136000-25G
BT136000-100G
BT136000-500G
Dimethyl acetylenedicarboxylate ≥98%
Dimethyl acetylenedicarboxylate
- RTECS: ES0175000
- Refractive index: 1.447
- Light Sensitive
- Liquid
Formula:
C₆H₆O₄ MW: 142.11 g/mol Boiling Pt: 95…98 °C/19 mmHg Density: 1.156 g/mL Flash Pt: 86 °C (187 °F) |
MDL Number:
MFCD00008456 CAS Number: 762-42-5 EINECS: 212-098-4 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...