BEAN135620

135620-1KG 135620-250G
BT135620-1KGEA 126.11 USD
BT135620-1KG BT135620-250G
BEAN135620
  • RTECS: KH9275000
  • Refractive index: 1.539
  • Light Sensitive
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR