1-Chlorododecane ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Lauryl chlorideDodecylchloride

135600-10ML
BT135600-10MLEA 70.32 USD
BT135600-10ML
1-Chlorododecane ≥99.5%
1-Chlorododecane

  • Refractive index: 1.433
  • Liquid

Formula: C₁₂H₂₅Cl
MW: 204.78 g/mol
Boiling Pt: 260 °C
Melting Pt: –9.3 °C
Density: 0.867 g/mL
Flash Pt: 130 °C (266 °F)
MDL Number: MFCD00013688
CAS Number: 112-52-7
EINECS: 203-981-5
ADR: 9,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR