BEAN134710

134710-5G 134710-25G
BT134710-5GEA 78.64 USD
BT134710-5G BT134710-25G
BEAN134710
  • RTECS: NT8060000
  • Refractive index: 1.464
  • Moisture Sensitive
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR