Methyl diphenyl phosphite
Supplier: BeanTown Chemical
134440-5G
134440-1G
134440-25G
BT134440-1GEA
58.07
USD
BT134440-1G
BT134440-5G
BT134440-25G
Methyl diphenyl phosphite
Methyl diphenyl phosphite
- Hygroscopic
- Liquid
Formula:
C₁₃H₁₃O₃P |
MDL Number:
MFCD00014886 UN: 3278 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...