Diethyl methylphosphonite

Supplier: BeanTown Chemical

134385-2G 134385-500MG 134385-10G
BT134385-500MGEA 50.45 USD
BT134385-500MG BT134385-10G BT134385-2G
Diethyl methylphosphonite
Diethyl methylphosphonite

  • Refractive index: 1.419
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₅H₁₃O₂P
Density: 0.907 g/mL
Flash Pt: 29 °C (84 °F)
MDL Number: MFCD00009089
CAS Number: 15715-41-0
EINECS: 239-805-9
UN: 3279
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR