1,3,5-Tris(bromomethyl)benzene ≥97%
Supplier: BeanTown Chemical
134310-5G
BT134310-5GEA
117.18
USD
BT134310-5G
1,3,5-Tris(bromomethyl)benzene ≥97%
1,3,5-Tris(bromomethyl)benzene
- Solid
Formula:
C₉H₉Br₃ Melting Pt: 94…99 °C |
MDL Number:
MFCD00182500 CAS Number: 18226-42-1 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...