Phenanthrene ≥98%

Supplier: BeanTown Chemical

134275-250G 134275-50G
BT134275-250GEA 201.46 USD
BT134275-250G BT134275-50G
Phenanthrene ≥98%
Phenanthrene

  • RTECS: SF7175000
  • Solid

Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340 °C
Melting Pt: 98…100 °C
Density: 1.063 g/mL
Flash Pt: 171 °C (339 °F)
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/84/56/2298456.jpg

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR