Cacodylic acid sodium salt trihydrate

Synonyms: Dimethylarsonic acid sodium salt trihydrateSodium cacodylate trihydrateSodium dimethylarsinate trihydrateDimethylarsinic acid sodium salt trihydrate

133625-25G 133625-100G
BT133625-25GEA 65.5 USD
BT133625-25G BT133625-100G
Cacodylic acid sodium salt trihydrate
Cacodylic acid sodium salt trihydrate

  • RTECS: CH7700000
  • Crystalline/Powder

Formula: (CH₃)₂As(O)ONa·3H₂O
MW: 213.94 g/mol
MDL Number: MFCD00149079
CAS Number: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR