4-Bromo-1-butene ≥97%
Supplier: BeanTown Chemical
133520-5G
133520-25G
BT133520-5GEA
63.67
USD
BT133520-5G
BT133520-25G
4-Bromo-1-butene ≥97%
4-Bromo-1-butene
- Refractive index: 1.462
- Light Sensitive
- Liquid
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 98…100 °C Density: 1.33 g/mL Flash Pt: 1 °C (34 °F) |
MDL Number:
MFCD00000258 CAS Number: 5162-44-7 EINECS: 225-937-4 UN: 1993 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...