3-Buten-1-ol ≥98%
Supplier: BeanTown Chemical
Synonyms:
But-3-en-1-ol
, 4-Hydroxy-1-butene
133205-5G
133205-25G
BT133205-5GEA
44.25
USD
BT133205-5G
BT133205-25G
3-Buten-1-ol ≥98%
3-Buten-1-ol
- Refractive index: 1.421
- Liquid
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 112…114 °C Density: 0.838 g/mL Flash Pt: 33 °C (91 °F) |
MDL Number:
MFCD00002959 CAS Number: 627-27-0 EINECS: 210-991-3 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...