Niobium carbide ≥99% (trace metals basis)
Supplier: BeanTown Chemical
132985-25G
132985-100G
BT132985-100GEA
132.93
USD
BT132985-100G
BT132985-25G
Niobium carbide ≥99% (trace metals basis)
Niobium carbide
- <10 Micron Powder
Formula:
CNb Melting Pt: 3500 °C Density: 7.6 g/mL |
MDL Number:
MFCD00016237 EINECS: 235-117-8 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...