4-Methylimidazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Methyl-1H-imidazole

132940-10G 132940-50G
BT132940-10GEA 35.58 USD
BT132940-10G BT132940-50G
4-Methylimidazole ≥98%
4-Methylimidazole

  • RTECS: NI7350000
  • Powder

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 263 °C
Melting Pt: 44…47 °C
Density: 1.020 g/mL
Flash Pt: 157 °C (315 °F)
MDL Number: MFCD00005201
CAS Number: 822-36-6
EINECS: 212-497-3
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR