BEAN132935

132935-100G 132935-5G 132935-25G
BT132935-5GEA 24.19 USD
BT132935-5G BT132935-25G BT132935-100G
BEAN132935
CAS: 506-68-3; EC No: 208-051-2; MDL No: MFCD00011597; RTECS: GT2100000
UN No: UN1889; Haz Class: 6.1 (8); Packing Group: I
Crystalline; Linear Formula: BrCN; MW: 105.93
Melting Point: 50-53°; Boiling Point: 61-62°
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR