BEAN132750

132750-5G 132750-25G
BT132750-5GEA 12.77 USD
BT132750-5G BT132750-25G
BEAN132750
CAS: 4039-32-1; EC No: 223-725-6; MDL No: MFCD00008261
UN No: UN2925; Haz Class: 4.1 (8); Packing Group: II
Powder; Molecular Formula: C6H18LiNSi2; MW: 167.33
Melting Point: 83-86°
Density (g/mL): 0.86
Air Sensitive, Moisture Sensitive
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR