N,N'-Diethylethylenediamine ≥96%
Supplier: BeanTown Chemical
132745-5G
BT132745-5GEA
83.09
USD
BT132745-5G
N,N'-Diethylethylenediamine ≥96%
N,N'-Diethylethylenediamine
- Refractive index: 1.433
- Liquid
Formula:
C₆H₁₆N₂ MW: 116.21 g/mol Boiling Pt: 152…154 °C Density: 0.811 g/mL Flash Pt: 33 °C (91 °F) |
MDL Number:
MFCD00009033 CAS Number: 111-74-0 EINECS: 203-902-4 UN: 2685 ADR: 8,II |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...