Hexythiazox

Supplier: BeanTown Chemical

Synonyms: rel-(4R,5R)-5-(4-Chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-3-thiazolidinecarboxamide

131385-100MG
BT131385-100MGEA 74.58 USD
BT131385-100MG
Hexythiazox
Hexythiazox

  • RTECS: XJ5396000
  • Solid

Formula: C₁₇H₂₁ClN₂O₂S
MW: 352.877 g/mol
Melting Pt: 106…109 °C
MDL Number: MFCD00137385
CAS Number: 78587-05-0
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR