N,N-Dimethylaniline ≥99%

Supplier: BeanTown Chemical

131250-500ML 131250-100ML
BT131250-100MLEA 25.82 USD
BT131250-100ML BT131250-500ML
N,N-Dimethylaniline ≥99%
N,N-Dimethylaniline

  • RTECS: BX4725000
  • Refractive index: 1.557
  • Liquid

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 193…194 °C
Melting Pt: 1.5…2.5 °C
Density: 0.956 g/mL
Flash Pt: 75 °C (167 °F)
MDL Number: MFCD00008304
CAS Number: 121-69-7
EINECS: 204-493-5
UN: 2253
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR