Potassium pyroantimonate ≥94%
Supplier: BeanTown Chemical
Synonyms:
Potassiumhexahydroxoantimonate(V)
, Potassium hexahydroxoantimonate
, Potassium antimonate
, Potassium antimonate trihydrate
131035-250G
131035-50G
BT131035-50GEA
110.64
USD
BT131035-50G
BT131035-250G
Potassium pyroantimonate ≥94%
Potassium pyroantimonate
- Powder
Formula:
KSbO₃·3H₂O MW: 262.9 g/mol |
MDL Number:
MFCD00011415 CAS Number: 12208-13-8 EINECS: 235-387-7 UN: 1549 ADR: 6.1,III |
Specification Test Results
Assay | 94% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...