Cyclopentane ≥99%
Supplier: BeanTown Chemical
Synonyms:
Pentamethylene
130685-4L
130685-250ML
130685-1L
BT130685-1LEA
141.42
USD
BT130685-1L
BT130685-250ML
BT130685-4L
Cyclopentane ≥99%
Cyclopentane
- RTECS: GY2390000
- Refractive index: 1.405
- Liquid
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 50 °C Melting Pt: –94 °C Density: 0.751 g/mL Flash Pt: –20 °C (–4 °F) |
MDL Number:
MFCD00001356 CAS Number: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...