4-Chloro-α,α,α-trifluorotoluene ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Chlorobenzotrifluoride1-Chloro-4-(trifluoromethyl)benzene

130625-100G 130625-2.5KG 130625-500G
BT130625-100GEA 22.34 USD InStock
BT130625-100G BT130625-2.5KG BT130625-500G
4-Chloro-α,α,α-trifluorotoluene ≥98%
4-Chloro-α,α,α-trifluorotoluene

CAS: 98-56-6; EC No: 202-681-1; MDL No: MFCD00000627; RTECS: XS9145000
UN No: UN2234; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C7H4ClF3; MW: 180.55
Melting Point: -36°; Boiling Point: 136-138°; Flash point: 43°C (109°F)
Density (g/mL): 1.353; Refractive Index: 1.446

Formula: C₇H₄ClF₃
MW: 180,56 g/mol
Boiling Pt: 136 °C (1013 hPa)
Melting Pt: -36 °C
Density: 1.346 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00000627
CAS Number: 98-56-6
EINECS: 202-681-1
UN: 2234
ADR: 3,III
Merck Index: 13,02145

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR