1,3-Dichloroacetone ≥96%

130515-500G 130515-25G 130515-100G
BT130515-500GEA 544.74 USD
BT130515-500G BT130515-100G BT130515-25G
1,3-Dichloroacetone ≥96%
1,3-Dichloroacetone

  • RTECS: UC1430000
  • Solid

Formula: ClH₂CC(O)CH₂Cl
MW: 126.97 g/mol
Boiling Pt: 173 °C
Melting Pt: 39…41 °C
Density: 1.383 g/mL
Flash Pt: 89 °C (192 °F)
MDL Number: MFCD00000937
CAS Number: 534-07-6
EINECS: 208-585-6
UN: 2649
ADR: 6.1,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR