2,2-Dimethylbutyryl chloride ≥98%

Synonyms: 2,2-Dimethylbutyrylchloride

130485-10G 130485-250G 130485-50G
BT130485-50GEA 90.78 USD
BT130485-50G BT130485-10G BT130485-250G
2,2-Dimethylbutyryl chloride ≥98%
2,2-Dimethylbutyryl chloride

  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Density: 0.98 g/mL
Flash Pt: 29 °C (84 °F)
MDL Number: MFCD03093071
CAS Number: 5856-77-9
EINECS: 227-478-5
ADR: 8 (3),II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR