Cadmium perchlorate hexahydrate ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

130355-25G
BT130355-25GEA 142.28 USD
BT130355-25G
Cadmium perchlorate hexahydrate ≥99.999% (trace metals basis)
Cadmium perchlorate hexahydrate

  • Hygroscopic
  • Crystalline

Formula: CdCl₂O₈·6H₂O
MW: 419.4 g/mol
MDL Number: MFCD00015992
CAS Number: 10326-28-0
UN: 1481
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR