Glyoxylic acid 50% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: 2-Oxoacetic acid

129885-5KG 129885-250G 129885-1KG
BT129885-5KGEA 384.16 USD
BT129885-5KG BT129885-250G BT129885-1KG
Glyoxylic acid 50% (w/w) in water
Glyoxylic acid

  • Liquid

Formula: C₂H₂O₃
MW: 74.04 g/mol
Density: 1.342 g/mL
MDL Number: MFCD00006958
EINECS: 206-058-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR