Diisopropylamine ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-Isopropyl-2-propanamine
129560-2.5L
129560-500ML
BT129560-2.5LEA
102.18
USD
BT129560-2.5L
BT129560-500ML
Diisopropylamine ≥99%
Diisopropylamine
- RTECS: IM4025000
- Refractive index: 1.392
- Liquid
Formula:
C₆H₁₅N MW: 101.19 g/mol Boiling Pt: 84 °C Melting Pt: –61 °C Density: 0.722 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00008862 CAS Number: 108-18-9 EINECS: 203-558-5 UN: 1158 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...