BEAN129165

129165-100G 129165-25G
BT129165-100GEA 160.26 USD
BT129165-100G BT129165-25G
BEAN129165
  • RTECS: KN3950000
  • Refractive index: 1.445
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR