Diarsenic triselenide ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Arsenic(III) selenide
128670-25G
128670-500MG
128670-5G
BT128670-500MGEA
32
USD
BT128670-500MG
BT128670-5G
BT128670-25G
Diarsenic triselenide ≥99.999% (trace metals basis)
Diarsenic triselenide
- RTECS: CG2285000
- 1-6mm Fused Pieces
Formula:
As₂Se₃ MW: 386.72 g/mol |
MDL Number:
MFCD00014166 CAS Number: 1303-36-2 EINECS: 215-119-5 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...