Cerium trinitrate hexahydrate ≥99.99% (trace rare earth metals basis)

Synonyms: Cerium (III) nitrate hexahydrateCerous chloride hexahydrate

128635-250G
BT128635-250GEA 720.81 USD
BT128635-250G
Cerium trinitrate hexahydrate ≥99.99% (trace rare earth metals basis)
Cerium trinitrate hexahydrate

  • RTECS: FK6300000
  • Hygroscopic
  • Crystalline Aggregates

Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR