2,6-Xylidine ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,6-Dimethylaniline2-Amino-m-xylene

128565-1KG 128565-100G 128565-500G
BT128565-1KGEA 146.41 USD
BT128565-1KG BT128565-500G BT128565-100G
2,6-Xylidine ≥99%
2,6-Xylidine

  • RTECS: ZE9275000
  • Refractive index: 1.560
  • Liquid

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 214 °C/739 mmHg
Melting Pt: 10…12 °C
Density: 0.984 g/mL
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00007747
CAS Number: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR